Learn More
Brady™ BradyPrinter i3300 Rotating Self-laminating Vinyl Wire Labels
Identify your wires from any angle with B-427 Rotating Vinyl Wire Labels. Simply print your labels and then peel, fold, and wrap – they’ll rotate freely for easy ID on even closed network connections.
Supplier:  Brady™ B33RO295427
 View more versions of this product
Catalog No. 19129516
Only null left
Description
Description
  • Rotates so you can identify your labels from any angle
  • Simply wrap and apply – there is no adhesive residue or label parts to cling to your wires
  • Spins freely without anchors so these labels won't damage your Cat 5/6 and fiber optic cables
Specifications
Chemical Identifiers
Wire Labels
BradyPrinter i3300
Permanent
Rectangle
Wire and Cable Labeling
White/Translucent
Self-laminating Vinyl
Matte
-94°F to +158°F
Specifications
Wire Labels
BradyPrinter i3300
1 in.
10 to 6 AWG
Matte
-94°F to +158°F
700/Pk.
White/Translucent
Self-laminating Vinyl
2.75 in.
Permanent
Rectangle
Wire and Cable Labeling
2
Product Suggestions
Product Suggestions