Learn More
Brady™ Oil Plus™ Heavy Pad, Dispense-n-Dispose
Catalog No. 296353202
Change view
Click to view available options
Capacity (English):
39 gal./Bale
Catalog No. Capacity (English)
296353202 39 gal./Bale
1 options
Catalog No. 296353202 Supplier Brady™ Supplier No. OP150DND
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Oil Plus™ Dispense-n-Dispose heavy pads absorb petroleum-based oils, solvents and other non-water soluble liquids.
  • White absorbents pick up petroleum-based fluids, paints and non-water soluble chemicals while repelling water - great for indoor and outdoor use
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • 3-ply meltblown polypropylene construction provides durability; pads float indefinitely even when saturated with oil
  • Perforations provide cost control and reduce waste by allowing you to use only what you need
Specifications
Color White
Capacity (English) 39 gal./Bale
Material Polypropylene
Type Oil Plus Heavy Pad, Dispense-n-Dispose
For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up, drums - liquid storage, outfalls and discharge ponds, heavy manufacturing, indoor/outdoor petroleum clean-up
Packaging Quantity 150/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 38.1 x 48.2 cm (15 x 19 in.)
Length (Metric) 48.26 cm
Width (Metric) 38.1 cm