Learn More
Brady™ Labels for Desktop Laser Printers
Labels for Desktop Laser Printers ideal for rating plates, asset identification, and general purpose labeling applications.
Supplier:  Brady™ LAT607992.5
 Afficher plus de versions de ce produit
Numéro de catalogue. 11877153
Il en reste null
Description
Description
  • Durable, chemical- and solvent-resistant labels for use with a variety of laser printers.
  • High resolution, good solvent and smudge resistance, moderate to high temperature performance.
  • Withstand extreme laboratory environments.
Spécifications
Identifiants chimiques
Autoclavable
Acrylic, Permanent
Label
Non-sterile
LaserTab Laser Printable Labels
Matte
Laser
Spécifications
Autoclavable
LaserTab Laser Printable Labels
White
Matte
Nylon Cloth
Laser
Non-sterile
Laser Printers
Pack of 9 Sheets
-40°C
2500 Pk.
0.375 in. dia.
-40°C to +90°C
0.165 mm
0.375 in.
9.52 mm
0.37 in.
LAT-60-799-2.5
Halogen Free (DIN VDE 0472/part 815)
Acrylic, Permanent
Consignment
Label
Single-side Printable
Label
Freezers, Liquid Nitrogen, Autoclaves, Hot Water Baths, Laboratory Labeling and Identification
12.70 mm
15
2565 Labels/Pack
Round
-40°F to +194°F
6.5 mil
0.375 x 0.375 in. (9.35 x 9.35 mm)
0.375 in.
12.70 mm
0.952 cm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS