missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazwik 65 gal. Lab Pack Kit

Numéro de catalogue. 19000194
Modifier l'affichage
Click to view available options
Capacity (English):
65 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacity (English)
19000194 65 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 19000194 Fournisseur Brady™ Code fournisseur SKH65
Il en reste null

Comprend

100 Pads, 7 Socs, 7 Pillows, 1 Pair Gloves, 5 Disposal Bags, 1 Pair Goggles, EmergencyResponse Handbook

Hazwik™ 65 gal. Drum Spill Kit provides a fast response to medium-large sized spills of hazardous chemical materials.

  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • 65-Gallon Drum is a UN-approved, tough, secure, screw-topped drum ideal for medium to large spill response
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents

Spécifications

Color Yellow
Capacity (English) 65 gal.
Material Polyethylene
Type Allwik Lab Pack Spill Kit
For Use With (Application) Medium-large spill responses
Includes 100 pads, 7 SOCs, 7 pillows, 1 pair gloves, 5 disposal bags, 1 pair goggles, emergency response handbook
Height (English) 33.5 in.
Diameter (English) 31 in.
Weight (English) 99.5 lb.
Weight (Metric) 219.4 kg