Learn More
Brady™ GlobalMark™ Polyester Tapes
Durable outdoor-grade polyester with permanent adhesive.
Supplier:  Brady™ 76699
 Afficher plus de versions de ce produit
Numéro de catalogue. 19201182
Il en reste null
Description
Description
  • Low-halide content; ideal for marking stainless-steel pipes and surfaces
  • Ultrasmooth finish is excellent for mounting to rigid panels and very smooth surfaces
  • Two-year average outdoor durability in temperatures from 110°C to -40°C (230°F to -40°F); can be applied in temperatures as low as -18°C (0°F)
Order Info
Thickness: 3mil
Spécifications
Identifiants chimiques
Tape
Ideal for Stainless Steel and surfaces
-40°C to +110°C
Non-sterile
-40°F to 230°F
Spécifications
Repositionable
Consignment
Tape
Single-side Printable
Label Tape
Ideal for Stainless Steel and surfaces
2 Years
-40°F to 230°F
3 mil
100 ft.
1.125 in.
White
Gloss
Vinyl
Thermal Transfer
Non-sterile
GlobalMark™ 2
1 Ea.
-40°C to +110°C
100 ft. x 1.125 in.
30.48 m
28.57 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS