missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Globally Harmonized System (GHS) Corrosive Picto Labels

Globally Harmonized System (GHS) Corrosive Picto Labels for corrosives.

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance GHS, OSHA 29 CFR 1910.1200
Description GHS Corrosive Labels
Color Red, Black, White
Format Label
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Material Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Material Price Quantity & Availability  
19140516
View Documents Promotion Details
Brady™
118839
39 Pk. Vinyl N/A
Only null left
 
19140525
View Documents Promotion Details
Brady™
118893
39 Pk. Paper N/A
Only null left
 
19140534
View Documents Promotion Details
Brady™
118840
16 Pk. Vinyl N/A
Only null left
 
19140543
View Documents Promotion Details
Brady™
118894
16 Pk. Paper N/A
Only null left
 
19140703
View Documents Promotion Details
Brady™
118841
4/Card Vinyl (B-7569) N/A
Only null left
 
Description

Description

For Corrosives

Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards

  • Legend: Corrosive
  • Made of B-7569 Vinyl or Paper
  • Available in three sizes
Specifications

Specifications

Not Autoclavable
GHS Corrosive Labels
Label
Corrosive
GHS, OSHA 29 CFR 1910.1200
Red, Black, White
Non-sterile
Videos
SDS
Documents

Documents