missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Corrosive Picto Labels
Globally Harmonized System (GHS) Corrosive Picto Labels for corrosives.
Supplier: Brady™ 118841
Description
For Corrosives
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Corrosive
- Made of B-7569 Vinyl or Paper
- Available in three sizes
Chemical Identifiers
| Not Autoclavable | |
| GHS Corrosive Labels | |
| Label | |
| Corrosive |
| GHS, OSHA 29 CFR 1910.1200 | |
| Red, Black, White | |
| Non-sterile |
Specifications
| Not Autoclavable | |
| GHS Corrosive Labels | |
| Red, Black, White | |
| Label | |
| Non-sterile | |
| General labeling | |
| 3 Years | |
| Diamond | |
| -4°F to +167°F | |
| 4 mil | |
| 2 x 2 in. | |
| 50.80 mm |
| GHS, OSHA 29 CFR 1910.1200 | |
| Permanent Acrylic | |
| Gloss | |
| Vinyl (B-7569) | |
| Corrosive | |
| Card of 4 Labels | |
| 4/Card | |
| 50.80 x 50.80 mm (L x W) | |
| -20°C to +75°C | |
| 0.102 mm | |
| 2 in. |