missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Corrosive Picto Labels
Globally Harmonized System (GHS) Corrosive Picto Labels for corrosives.
Supplier: Brady™ 118840
Description
For Corrosives
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Corrosive
- Made of B-7569 Vinyl or Paper
- Available in three sizes
Chemical Identifiers
| Not Autoclavable | |
| GHS Corrosive Labels | |
| Label | |
| Corrosive |
| GHS, OSHA 29 CFR 1910.1200 | |
| Red, Black, White | |
| Non-sterile |
Specifications
| Not Autoclavable | |
| GHS Corrosive Labels | |
| Red, Black, White | |
| Label | |
| Pictogram Label | |
| Corrosive | |
| Card of 16 Labels | |
| 16 Pk. | |
| 25.40 x 25.40 mm (L x W) | |
| -20°C to +75°C | |
| 0.102 mm | |
| 1 in. | |
| 1 in. |
| GHS, OSHA 29 CFR 1910.1200 | |
| Permanent Acrylic, Permanent | |
| Gloss | |
| Vinyl | |
| Non-sterile | |
| For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification | |
| 3 Years | |
| Diamond | |
| -4°F to +167°F | |
| 0.004 in. | |
| 1 x 1 in. | |
| 25.40 mm | |
| 25.40 mm |