missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Corrosive Picto Labels
Globally Harmonized System (GHS) Corrosive Picto Labels for corrosives.
Supplier: Brady™ 118894
Description
For Corrosives
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Corrosive
- Made of B-7569 Vinyl or Paper
- Available in three sizes
Identifiants chimiques
| Not Autoclavable | |
| GHS Corrosive Labels | |
| Label | |
| Corrosive |
| GHS, OSHA 29 CFR 1910.1200 | |
| White, Black, Red | |
| Non-sterile |
Spécifications
| Not Autoclavable | |
| GHS Corrosive Labels | |
| White, Black, Red | |
| Label | |
| Pictogram Label | |
| Corrosive | |
| Card of 16 Labels | |
| 16 Pk. | |
| 25.40 x 25.40 mm (L x W) | |
| -40°C to +65°C | |
| 0.097 mm | |
| 1 in. | |
| 1 in. |
| GHS, OSHA 29 CFR 1910.1200 | |
| Permanent Acrylic, Pressure-sensitive | |
| Semi-gloss | |
| Paper | |
| Non-sterile | |
| General labeling | |
| 2 Years | |
| Diamond | |
| -40°F to +150°F | |
| 0.0038 in. | |
| 1 x 1 in. | |
| 25.40 mm | |
| 25.40 mm |