missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: No Smoking Signs

Various sizes and materials

$20.40 - $47.33

Specifications

Certifications/Compliance OSHA
Quantity 1 Ea.
Min. Temperature (English) -40°F
Displayed Text DANGER No Smoking
Product Type No Smoking Sign
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19102050
View Documents Promotion Details
Brady™
25077
Plastic
Each for $20.40
Only null left
 
1704615
View Documents Promotion Details
Brady™
47010
Fiberglass
Each for $47.33
Only null left
 
Description

Description

AL: BradyTuff™ aluminum
FG: Premium fiberglass
PL: BradyTuff plastic
SS: Self-sticking high performance polyester

Specifications

Specifications

OSHA
-40°F
No Smoking Sign
1 Ea.
DANGER No Smoking
Videos
SDS
Documents

Documents