missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Chemical and Hazardous Materials Signs: DANGER - EXPLOSIVES
Identify equipment, rooms and materials which contain or are contaminated with hazardous agents
$13.59 - $46.60
Specifications
| Certifications/Compliance | ANSI Z535, OSHA 1910.145 |
|---|---|
| Quantity | 1 Ea. |
| Mounting Type | Surface Mount |
| For Use With (Application) | Designed for use in industrial, utility, commercial and institutional environments |
| Min. Temperature (English) | -40°F |
| Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Thickness | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Thickness | Price | Quantity | ||
|
19035933
|
Brady™
BRD25654 |
Plastic | 7 in. | 10 in. | 0.06 in. |
Each for $13.59
|
|
||
|
19-035-934
|
Brady™
25655 |
Plastic | 10 in. | 14 in. | 0.06 in. |
Each for $21.78
|
|
||
|
19-035-932
|
Brady™
75639 |
Fiberglass | 10 in. | 14 in. | 0.1 in. |
Each for $46.60
|
|
||
Description
- Available in three materials - plastic, fiberglass and polyester
- Available in different widths and heights
- Black/red on white
Specifications
| ANSI Z535, OSHA 1910.145 | |
| Surface Mount | |
| -40°F | |
| Hazardous Materials Sign |
| 1 Ea. | |
| Designed for use in industrial, utility, commercial and institutional environments | |
| Explosive |