missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Chemical and Hazardous Materials Signs: DANGER - EXPLOSIVES
Identify equipment, rooms and materials which contain or are contaminated with hazardous agents
Supplier: Brady™ 75639
Description
- Available in three materials - plastic, fiberglass and polyester
- Available in different widths and heights
- Black/red on white
Specifications
| ANSI Z535, OSHA 1910.145 | |
| 1 Ea. | |
| Surface Mount | |
| 10 in. | |
| 193°F | |
| 0.1 in. | |
| 35.56 cm | |
| -40°F to 193°F | |
| Hazardous Materials Sign |
| Fiberglass | |
| 9000 psi | |
| Designed for use in industrial, utility, commercial and institutional environments | |
| 25.40 cm | |
| -40°F | |
| 14 in. | |
| -40°C to 90°C | |
| Explosive | |
| Red on White |