missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Chemical and Hazardous Materials Signs: Asbestos

Catalog No. 19016965
Change view
Click to view available options
Color:
Black on Yellow
Red on White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19016965 Red on White
19016357 Black on Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19016965 Supplier Brady™ Supplier No. 74520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Brady chemical and hazardous materials signs are used to identify hazardous materials, like Asbestos, to help sustain a safe workplace.

  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145
Material Fiberglass
Quantity 1 Ea.
Tensile Strength 9000 psi
Adhesive Type Non-adhesive
Description Premium Fiberglass
Mounting Type Surface Mount
For Use With (Application) Industrial, utility, commercial and institutional environment
Height (English) 20 in.
Max. Temperature (English) 193°F
Min. Temperature (English) -40°F
Thickness 0.1 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 90°C
Temperature Range (English) -40°F to 193°F
Dimensions (L x W) 20 x 14 in.
Displayed Text DANGER Asbestos Cancer And Lung Disease Hazard Authorized Personnel Only Respirators And Protective Clothing are Required In This Area
Product Type Chemical and Hazardous Materials Sign
Color Red on White
Show More Show Less