missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Blank Hazardous Communication Panels
Description
Requires
Quick-Align™ numbers (0 through 4) and symbols (Oxygen, No Water, Radiation, Acid, Corrosive, and Alkali) needed to customize signs sold separately in sizes from 1 to 6 in. . Contact your Fisher Safety Customer Service Representative for ordering information.
Make signs to identify hazardous chemical containers and storage areas.
- Place directly on storage tanks, processing tanks and vessels, feed hoppers, pipelines, and other potentially hazardous equipment
- Upper three corners of signal rate health, fire and reactivity on a scale of 0 to 4; lower corner provides specific hazard information
- Available in vinyl with permanent pressure-sensitive adhesive or in rigid fiberglass with predrillled holes for mounting to walls
Compliance
Signals meet NFPA Code 704M and Federal Standard 313.
Specifications
Specifications
| Material | Fiberglass |
| Quantity | 1 Ea. |
| Description | Premium Fiberglass |
| Mounting Type | Corner Holes |
| Height (English) | 11 in. |
| Width (English) | 11 in. |
| Shape | Square |
| Languages | English |
| Displayed Text | NFPA Diamond |
| Product Type | Blank Sign |
| Show More |