Learn More
Brady™ Admittance Signs: Caution Do Not Enter with Pictogram (ANSI Z535)
Admittance Signs: Caution: Do Not Enter with Pictogram (ANSI Z535) are safety signs from Brady™ that help to manage and sustain a clean and safe workplace. They are available in a variety of materials and sizes that best fit your application.
$20.40 - $38.08
Specifications
| Quantity | 1 Ea. |
|---|---|
| Adhesive Type | Non-adhesive |
| Min. Temperature (English) | -40°F |
| Displayed Text | CAUTION Do Not Enter |
| Product Type | Admittance Sign |
Description
- Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
- Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
- Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
| 1 Ea. | |
| -40°F | |
| Admittance Sign |
| Non-adhesive | |
| CAUTION Do Not Enter |