missing translation for 'onlineSavingsMsg'
Learn More

Brady™ 30 gal. Spill Kit Refill, Chemical

Catalog No. 17111234
Change view
Click to view available options
Capacity (English):
30 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17111234 30 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17111234 Supplier Brady™ Supplier No. SKH30R

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Spill Kit Refill is intended for use with the Hazwik™ 30 gal. chemical spill kit. (Refill kit only—container sold separately.)

  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents

Specifications

Color Green
Capacity (English) 30 gal.
Capacity (Metric) 113.5 L
Type Chemical Spill Kit Refill
For Use With (Application) Chemical spills
Includes (25) 15 x 19 in. Pads, (6) 3 in. x 12 ft. socks, (6) 18 x 18 in. pillows, 1 pair goggles, 1 pair Nitrile gloves, 3 disposal bags, handbook