Learn More
Microwave Research & Applications Inc BP110 Laboratory Grade Microwave, 120V, 20 w x 12 1/8 h x 16 d (W/O handle and vent duct)
Numéro de catalogue. NC9528110
Encompass
The BP-110 is a laboratory grade microwave with time and power based controls much like a domestic microwave oven that many laboratories use every day. It produces up to 1,000 watts of microwave power, the most common power level of domestic microwaves sold today meaning you should be able to use your existing protocol. The microwave has a powered cavity ventilation system that can be connected into the house air handling system or to an approved laboratory air handling system. Cavity 330mm wide x 205mm tall x 330mm deep (13 x 8 1/16 x 13) Operates just as a domestic microwave oven using time and power level120 volt operation, 15 ampere needs no special wiring (230 VAC 50 Hertz input available)Up to 1,000 watts of microwave power.
Numéro de catalogue. NC9528110 Fournisseur Microwave Research & Applications Inc Code fournisseur BP110
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The BP-110 is a laboratory grade microwave with time and power based controls much like a domestic microwave oven that many laboratories use every day. It produces up to 1,000 watts of microwave power, the most common power level of domestic microwaves sold today meaning you should be able to use your existing protocol. The microwave has a powered cavity ventilation system that can be connected into the house air handling system or to an approved laboratory air handling system. Cavity 330mm wide x 205mm tall x 330mm deep (13 x 8 1/16 x 13) Operates just as a domestic microwave oven using time and power level120 volt operation, 15 ampere needs no special wiring (230 VAC 50 Hertz input available)Up to 1,000 watts of microwave power.