missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine SPARC Native Protein

Catalog No. PIRP43148
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43148 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43148 Supplier Invitrogen™ Supplier No. RP43148

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43148 contains native bovine Osteonectin protein. RP-43148 may be used as a positive control in many immunological assays.

SPARC gene encodes a cysteine-rich acidic matrix-associated protein. The encoded protein is required for the collagen in bone to become calcified but is also involved in extracellular matrix synthesis and promotion of changes to cell shape. The gene product has been associated with tumor suppression but has also been correlated with metastasis based on changes to cell shape which can promote tumor cell invasion. Three transcript variants encoding different isoforms have been found for this gene.

Specifications

Accession Number P13213
For Use With (Application) Neutralization
Formulation TBS with 0.15M NaCl and no preservative; pH 7.4
Gene ID (Entrez) 282077
Molecular Weight (g/mol) 29 kDa
Name Bovine SPARC
pH Range 7.4
Purification Method SDS-PAGE
Quantity 50 μg
Source Bovine
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Basement membrane protein 40; basement-membrane protein 40; BM40; BM-40; Cysteine rich protein; OI17; ON; Osteonectin; osteonectin precursor; Secreted acidic cystein rich glycoprotein; secreted acidic cysteine rich glycoprotein; Secreted acidic cystein-rich glycoprotein (osteonectin); secreted protein acidic and cysteine rich; secreted protein acidic and rich in cysteine; Secreted protein acidic and rich in cysteine (SPARC); secreted protein, acidic, cysteine-rich (osteonectin); secreted protein, acidic, cysteine-rich [osteonectin]; SPARC
Common Name SPARC
Gene Symbol SPARC
Product Type Protein
Conjugate Unconjugated
Species Bovine
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less