missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Factor Va Native Protein

Catalog No. PIRP43129
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43129 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43129 Supplier Invitrogen™ Supplier No. RP43129

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43129 contains native bovine Factor Va protein. RP-43129 may be used as a positive control in many immunological assays.

Specifications

Accession Number Q28107
Concentration 2.4 mg/mL
For Use With (Application) Bioactivity
Formulation Protein with 5mM CaCl2, 50% glycerol and no preservative
Gene ID (Entrez) 280687
Molecular Weight (g/mol) 168 kDa
Name Bovine Factor Va
Purification Method SDS-PAGE
Quantity 100 μg
Source Bovine
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activated protein C cofactor; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; F5a; factor V; factor V Leiden; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2
Common Name Factor Va
Gene Symbol F5
Biological Activity Specific Activity = 1792 U/mg; determined by Factor V clotting assay where 1 U equals the Factor V activity in 1 mL of normal human plasma.
Product Type Protein
Conjugate Unconjugated
Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Specific Activity = 1792 U/mg; determined by Factor V clotting assay where 1 U equals the Factor V activity in 1 mL of normal human plasma.
Form Liquid
Protein Subtype Native
Show More Show Less