missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric anhydride, 99.98% trace metals basis, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 339075500G
Specifications
| 450°C | |
| 500 g | |
| MFCD00011315 | |
| 69.62 | |
| Poly Bottle |
| 1303-86-2 | |
| B2O3 | |
| Boron trioxide | |
| 99.98% | |
| 2.46 g/mL (at 25°C); Vapor Density: >1 (vs air) |
Chemical Identifiers
| MFCD00011315 | |
| Boron trioxide |