missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.5-100.5%, powder, Honeywell Fluka™
Puriss., meets analytical specification of BP, NF, Ph. Eur., 99.5-100.5%, powder
$477.38 - $2520.40
Chemical Identifiers
| CAS | 10043-35-3 |
|---|---|
| Molecular Formula | BH3O3 |
| Molecular Weight (g/mol) | 61.83 |
| MDL Number | MFCD00011337 |
| InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
| Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
| PubChem CID | 7628 |
| ChEBI | CHEBI:33118 |
| IUPAC Name | boric acid |
| SMILES | OB(O)O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 10043-35-3 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| 7628 | |
| boric acid |
| BH3O3 | |
| MFCD00011337 | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
| CHEBI:33118 | |
| OB(O)O |
Specifications
| 10043-35-3 | |
| 1 kg | |
| H3BO3 | |
| NONH for all modes of transport | |
| In accordance with ethanol | |
| OB(O)O | |
| 61.83 | |
| CHEBI:33118 | |
| 99.5 to 100.5% | |
| Boric Acid, Meets analytical specification of Ph. Eur.BP, NF |
| 2.6mmHg (20°C) | |
| BH3O3 | |
| MFCD00011337 | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 7628 | |
| 61.83g/mol | |
| puriss. |
Safety and Handling
P201-P308 + P313
H360FD
EINECSNumber : 233-139-2
RTECSNumber : ED4550000