missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.5-100.5%, powder, Honeywell Fluka™

Puriss., meets analytical specification of BP, NF, Ph. Eur., 99.5-100.5%, powder

Supplier:  Honeywell Chemicals 1160725KG

 View more versions of this product

Catalog No. 6001909


Only null left
Explore available promotions

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Boric Acid
10043-35-3
2.5 kg
H3BO3
NONH for all modes of transport
In accordance with ethanol
OB(O)O
61.83
CHEBI:33118
99.5 to 100.5%
Meets analytical specification of Ph. Eur.BP, NF
2.6mmHg (20°C)
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83g/mol
puriss.

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
OB(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P308 + P313

H360FD

EINECSNumber : 233-139-2

RTECSNumber : ED4550000

SDS
Documents

Documents

Promotions

Promotions