missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.5-100.5%, powder, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécifications
Spécifications
| Vapor Pressure | 2.6mmHg (20°C) |
| Quantity | 6 x 1 kg |
| Linear Formula | H3BO3 |
| UN Number | NONH for all modes of transport |
| Solubility Information | In accordance with ethanol |
| Formula Weight | 61.83 |
| Percent Purity | 99.5 to 100.5% |
| Grade | puriss. |
| Chemical Name or Material | Boric Acid, Meets analytical specification of Ph. Eur.BP, NF |
Sécurité et manipulation
| Hazard Statement | P201-P308 + P313 |
| Precautionary Statement | H360FD |
| EINECSNumber | 233-139-2 |
| RTECSNumber | ED4550000 |