missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Powder, NF, 99.5-100.5%, Spectrum™ Chemical
BO110, 10043-35-3, H3BO3
Supplier: Spectrum Chemical Mfg Cor BO110500GM
Description
Spectrum™ Chemical Boric Acid, Powder, NF is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..Specifications
| 10043-35-3 | |
| Poly Bottle | |
| BH3O3 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 99.5 to 100.5% |
| 100% | |
| 500 g | |
| MFCD00011337 | |
| OB(O)O | |
| 61.83 | |
| NF |
Chemical Identifiers
| 10043-35-3 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| OB(O)O |
| BH3O3 | |
| MFCD00011337 | |
| boric acid |