Learn More
Boric acid, 99+%
Description
Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.
Solubility
Soluble in water and alcohols. Slightly soluble in acetone and pyridine.
Notes
Store in cool place. Moisture sensitive. Incompatible with potassium and acid anhydrides.
Specifications
Specifications
| pH | 3.8 to 4.8 |
| Melting Point | ∼185°C (decomposition) |
| Density | 1.435 |
| Odor | Odorless |
| Flash Point | None |
| Linear Formula | H3BO3 |
| Quantity | 500 g |
| Beilstein | 1697939 |
| Merck Index | 14,1336 |
| Solubility Information | Soluble in water and alcohols. Slightly soluble in acetone and pyridine. |
| Show More |
Safety and Handling
| Hazard Category | H360FD |
| Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
| Precautionary Statement | P201-P202-P260-P281-P308+P313-P501c |
| EINECSNumber | 233-139-2 |
| RTECSNumber | ED4550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only