missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, 99.99%, (trace metal basis), extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Lewis acid
Specifications
Specifications
| Color | White |
| Melting Point | 169.0°C |
| Packaging | Plastic bottle |
| Assay Percent Range | 99.99% |
| Linear Formula | H3BO3 |
| Quantity | 500 g |
| Physical Form | Crystals or Crystalline Powder |
| Fieser | 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70 |
| Merck Index | 15, 1339 |
| Solubility Information | Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol. |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS P Statement May damage fertility. May damage the unborn child. |
| Precautionary Statement | GHS P Statement: Immediately call a POISON CENTER or doctor/physician. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only