missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 99.6%, ACS reagent

Catalog No. AC423485000
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423485000 500 g Plastic bottle
AC423480020 2 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423485000 Supplier Thermo Scientific Chemicals Supplier No. 42348 5000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Brown-Red
Heavy Metals (as Pb) 10ppm max.
Non_volatile Matter 0.05 % max. (with MeOH)
Packaging Plastic bottle
Assay Percent Range 99.6%
Linear Formula H3BO3
Quantity 500 g
Physical Form Crystalline powder
Fieser 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
Merck Index 15, 1339
Formula Weight 61.83
Percent Purity ≥99.5% (Acid)
Grade ACS Reagent
Chemical Name or Material Boric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-139-2
RTECSNumber ED4550000
TSCA TSCA

RUO – Research Use Only