missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 4% (w/v), with Bromocresol Green-Methyl Red Indicator, Ricca Chemical

Catalog No. p-3251371
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
107032 1 L Natural Poly Bottle
10701 4 L Natural Poly Bottle
107025 10 L Cubitainer
10705 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 107032 Supplier Ricca Chemical Company Supplier No. RI107032
Only null left

For Analysis of Nitrogen (Kjeldahl)

Chemical Identifiers

CAS 7732-18-5 , 10043-35-3 , 62625-32-5 , 845-10-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Orange-Red
Concentration 4% (w/v)
CAS Min % 3.77, 94.0, 9.0E-4, 7.0E-4
CAS Max % 3.92, 98.0, 9.0E-4, 7.0E-4
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Percent Purity 4% W/V
Grade Laboratory
Chemical Name or Material Boric Acid, With Bromocresol Green-Methyl Red Indicator
Show More Show Less