missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | BH3O3 |
| Molecular Weight (g/mol) | 61.83 |
| MDL Number | MFCD00011337 |
| InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
| PubChem CID | 7628 |
| ChEBI | CHEBI:33118 |
| IUPAC Name | boric acid |
| SMILES | OB(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
106816
|
Ricca Chemical Company
106816 |
500 mL |
Each for $71.09
|
|
|||||
|
1068-32
|
Ricca Chemical Company
106832 |
1 L |
Each for $86.16
|
|
|||||
|
1068-1
|
Ricca Chemical Company
10681 |
4 L |
Each for $177.09
|
|
|||||
|
R106800020F
|
Ricca Chemical Company
R106800020F |
20 L |
Each for $847.69
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| CHEBI:33118 | |
| OB(O)O |
| BH3O3 | |
| MFCD00011337 | |
| 7628 | |
| boric acid |
Specifications
| Colorless | |
| 94.0,3.77 | |
| 98.0,3.93 | |
| Aqueous Solution | |
| BH3O3 | |
| MFCD00011337 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 7628 | |
| 4% W/V | |
| Boric Acid Solution |
| 7732-18-5,10043-35-3 | |
| 4% (w/v) | |
| Natural Poly Bottle | |
| 500 mL | |
| Liquid | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
| OB(O)O | |
| 61.83 | |
| CHEBI:33118 | |
| Laboratory |
Safety and Handling
ShelfLife : 24 months