missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 4% (w/v), Ricca Chemical

Catalog No. 106816
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
106816 500 mL
106832 1 L
10681 4 L
R106800020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 106816 Supplier Ricca Chemical Company Supplier No. 106816

Chemical Identifiers

CAS 7732-18-5 , 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Colorless
Concentration 4% (w/v)
CAS Min % 94.0, 3.77
CAS Max % 98.0, 3.93
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
Percent Purity 4% W/V
Grade Laboratory
Chemical Name or Material Boric Acid Solution
Show More Show Less