missing translation for 'onlineSavingsMsg'
Learn More

Borax, 1% (w/v), Ricca Chemical

Catalog No. 105016
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
105016 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 105016 Supplier Ricca Chemical Company Supplier No. 105016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1303-96-4 , 7732-18-5

Specifications

Color Colorless
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Percent Purity 1%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH ∼9.5
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 1% (w/v)
Chemical Name or Material Borax
Show More Show Less