missing translation for 'onlineSavingsMsg'
Learn More

Borane - tert-Butylamine Complex 95.0+%, TCI America™

Catalog No. B126425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B126425G 25 g
B1264100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B126425G Supplier TCI America Supplier No. B126425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7337-45-3
Molecular Formula C4H14BN
Molecular Weight (g/mol) 86.97
MDL Number MFCD00075635
InChI Key GKFJEDWZQZKYHV-UHFFFAOYSA-N
Synonym tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane
PubChem CID 6364547
IUPAC Name 2-methylpropan-2-amine borane
SMILES B.CC(C)(C)N

Specifications

Melting Point 99°C
Color White
Quantity 25 g
UN Number 2926
Formula Weight 86.97
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Borane - tert-Butylamine Complex
RTECSNumber EO3900000
TSCA Yes