missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borane dimethyl sulfide complex, 2M solution in THF, AcroSeal™
CAS: 13292-87-0 | C2H9BS | 75.96 g/mol
$146.68 - $146.68
Specifications
| Chemical Name or Material | Borane-methyl sulfide complex |
|---|---|
| Color | Colorless to Yellow |
| Name Note | 2M Solution in Tetrahydrofuran |
| CAS | 13292-87-0 , 109-99-9 , 75-18-3 |
| Molecular Formula | C2H9BS |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC185071000
|
Thermo Scientific Chemicals
185071000 |
100 mL | AcroSeal™ Glass bottle |
Each for $146.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
| Borane-methyl sulfide complex | |
| 2M Solution in Tetrahydrofuran | |
| C2H9BS | |
| MFCD00013189 | |
| 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50 | |
| 15, 1336 | |
| Solubility in water: reacts. Other solubilities: soluble in ethyl ether, tetrahydrofurane, hexane,, toluene, methylene chloride and other aprotic, solvents | |
| B.CSC | |
| 75.96 | |
| 75.95 | |
| −17°C |
| Colorless to Yellow | |
| 13292-87-0 , 109-99-9 , 75-18-3 | |
| (CH3)2S·BH3 | |
| 1.80 to 2.20M | |
| 0.8500g/mL | |
| dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
| RMHDLBZYPISZOI-UHFFFAOYSA-N | |
| (methylsulfanyl)methane borane | |
| 9833925 | |
| 0.85 | |
| Solution |
RUO – Research Use Only