accessibility menu, dialog, popup

UserName

H3CSCH3

CAS RN 75-18-3

IUPAC Name: (methylsulfanyl)methane
Synonyms: dimethyl sulfide|exact-S|dimethylsulfid|reduced-dmso|dimethyl sulfoxide(reduced)|sulfure de methyle|methylthiomethyl radical|dimethyl sulfide (natural)|methane, 1,1'-thiobis-|1,1'-thiobis-methane
Molecular Weight (g/mol): 62.13
Molecular Formula: C2H6S
InChi Key: QMMFVYPAHWMCMS-UHFFFAOYSA-N
SMILES: CSC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results