missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bolt™ Bis-Tris Plus XCell SureLock™ Mini-Cell Welcome Pack, 4-12%

Catalog No. NW0412F
Encompass
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
NW0412F 15-well
NW0412D 10-well
NW0412E 12-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NW0412F Supplier Thermo Scientific™ Supplier No. NW0412F

The Bolt Welcome Pack provides all of the necessary Bolt gels and buffers needed to begin using the XCell SureLock Mini-Cell.

The Bolt Welcome Pack provides all of the necessary Bolt gels and buffers needed to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.

The Welcome Pack contains:

  • XCell SureLock Mini-Cell (EI0001)
  • Bolt Bis-Tris Plus gels (2 boxes, 20 gels)
  • Bolt MES Running Buffer, 20X (B0002)
  • MES and MOPS Running Buffer, Powder (NP000105, NP000205)
  • Bolt LDS Sample Buffer, 4X (B0007)
  • Bolt Sample Reducing Agent, 10X (B0009)
  • PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the XCell SureLock Mini-Cell

The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.

About Bolt Bis-Tris Plus Gels

Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt gels to obtain optimal results for your day-to-day protein separation needs. Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8, 10, 12, and 4-12% gradient and multiple well formats. Bolt Bis-Tris gels feature wedge-shaped wells, which can easily load up to two times more sample volume allowing for detection of proteins in very dilute samples or visualization of low-abundance proteins.

For transferring your proteins to a membrane, we recommend using for Bolt Transfer Buffer (BT0006) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Specifications

Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Separation Range 3.5 to 260 kDa
Wells 15-well
Quantity 1 Welcome Pk.
Shipping Condition Approved for shipment at room temperature and wet ice
Product Line Bolt, WedgeWell
For Use With (Equipment) XCell SureLock Mini-Cell

For Research Use Only. Not for use in diagnostic procedures.