Learn More
Velp Scientifica™ BOD Sensor System 6
Catalog No. 07222011
Change view
Click to view available options
Voltage:
115 V
Catalog No. Voltage
07222011 115 V
1 options
Catalog No. 07222011 Supplier Velp Scientifica™ Supplier No. SA10210136
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Innovative, mercury-free and highly reliable solution for BOD analysis (Biochemical Oxygen Demand).
  • BOD Sensor System with 6 positions
  • Determination is a simple and easy way for monitoring microorganisms' activity in water samples with highly reliable results
  • Microorganisms present or inoculated in a water sample containing biodegradable organic substance, consume oxygen for their metabolic activity and produce a corresponding volume of carbon dioxide
  • Carbon dioxide gets absorbed by a strong alkali positioned in the neck of the bottle
  • Result of this process is a decrease in gas pressure
  • BOD measures this change in gas pressure between the beginning to the end of the analysis to give a result expressed in mg/L
Specifications
Type Sensor System
IP Rating IP54
Voltage 115 V
Certifications/Compliance CE