Learn More
Velp™ BOD Control Test Tablet
Catalog No. 11392026
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
11392026 10/Pk.
1 options
Catalog No. 11392026 Supplier Velp™ Supplier No. A00000136
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Velp™ BOD Measurement Test Tablet for use with BOD Sensors and BOD EVO Sensors.
  • For use with BOD and BOD EVO Sensors
Specifications
For Use With (Application) For verifying the full functionality of the RESPIROMETRIC Sensor System by simulating an oxygen demand experiment similar to the BOD method
Includes 10 Tablets/Box
Quantity 10/Pk.
Type BOD Control Test Tablet