missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC-Sarcosine, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Melting Point | 86°C to 90°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (ELSD) (HPLC) |
| Packaging | Glass bottle |
| Linear Formula | (CH3)3COCON(CH3)CH2COOH |
| Quantity | 5g, 5 g |
| Solubility Information | Solubility in water: slightly soluble |
| Formula Weight | 189.21 |
| Percent Purity | ≥99% |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| EINECSNumber | 237-306-0 |
| TSCA | TSCA |
RUO – Research Use Only