missing translation for 'onlineSavingsMsg'
Learn More

BOC-S-trityl-L-cysteine, 99%

Catalog No. AC399060050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399060050 5 g Glass Bottle
AC399060010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399060050 Supplier Thermo Scientific Chemicals Supplier No. 399060050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21947-98-8 | C27H29NO4S | 463.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21947-98-8
Molecular Formula C27H29NO4S
Molecular Weight (g/mol) 463.592
MDL Number MFCD00038251
InChI Key JDTOWOURWBDELG-QHCPKHFHSA-N
Synonym boc-cys trt-oh, n-boc-s-trityl-l-cysteine, boc-s-trityl-l-cysteine, n-tert-butoxycarbonyl-s-trityl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-cysteine trt-oh, 2r-2-tert-butoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-triphenylmethyl, boc-l-cysteine trityl, 2r-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid
PubChem CID 11167161
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

Assay Percent Range 99%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 463.6
Chemical Name or Material BOC-S-trityl-L-cysteine

RUO – Research Use Only