missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150196
Description
BOC Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
CDON and BOC are cell surface receptors of the immunoglobulin repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.Identifiants chimiques
| BOC | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6AZB0 | |
| 117606 | |
| Recombinant Brother Of CDO Homolog (BOC) (Lys668-Thr866). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Boc | |
| 4732455C11; biregional Cdon binding protein; biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein; BOC; BOC cell adhesion associated, oncogene regulated; Boc homolog; Boc homolog (mouse); brother of CDO; brother of CDON; CDON2; cell adhesion associated, oncogene regulated 2; mFLJ00376; Protein BOC; UNQ604/PRO1190 | |
| Boc | |
| Primary | |
| Protein A/G |
Spécifications
| BOC | |
| Polyclonal | |
| Unconjugated | |
| Boc | |
| 4732455C11; biregional Cdon binding protein; biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein; BOC; BOC cell adhesion associated, oncogene regulated; Boc homolog; Boc homolog (mouse); brother of CDO; brother of CDON; CDON2; cell adhesion associated, oncogene regulated 2; mFLJ00376; Protein BOC; UNQ604/PRO1190 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 117606 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6AZB0 | |
| Boc | |
| Recombinant Brother Of CDO Homolog (BOC) (Lys668-Thr866). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody |