missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Phenylalanine, 99+%

Catalog No. AC275641000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275641000 100 g Glass Bottle
AC275640050 5 g Glass Bottle
AC275640250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC275641000 Supplier Thermo Scientific Chemicals Supplier No. 275641000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 13734-34-4 | C14H19NO4 | 265.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13734-34-4
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00002663
InChI Key ZYJPUMXJBDHSIF-NSHDSACASA-N
Synonym boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
PubChem CID 77857
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 85.0°C to 88.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula C6H5CH2CH(COOH)NHCOOC(CH3)3
Specific Rotation + 25.00
Beilstein 14, 1595
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and methylene chloride, insoluble in aliphatic hydrocarbons
Water 0.5% max. (K.F.)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Specific Rotation Condition +25° (20°C c=1,ethanol)
Quantity 100 g
Formula Weight 265.3
Percent Purity ≥99%
Physical Form Fine Crystalline Powder
Chemical Name or Material BOC-L-Phenylalanine
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only