missing translation for 'onlineSavingsMsg'
Learn More

BOC-Glycine, 99+%

Catalog No. AC158801000
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158801000 100 g Glass Bottle
AC158800100 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158801000 Supplier Thermo Scientific Chemicals Supplier No. 158801000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4530-20-5 | C7H13NO4 | 175.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4530-20-5
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.18
MDL Number MFCD00002690
InChI Key VRPJIFMKZZEXLR-UHFFFAOYSA-N
Synonym boc-glycine, boc-gly-oh, n-tert-butoxycarbonyl glycine, n-boc-glycine, n-boc-l-glycine, tert-butoxycarbonylglycine, t-butoxycarbonylglycine, 2-tert-butoxycarbonyl amino acetic acid, glycine, n-1,1-dimethylethoxy carbonyl, tert-butyloxycarbonylglycine
PubChem CID 78288
SMILES CC(C)(C)OC(=O)NCC(O)=O

Specifications

Melting Point 87°C to 90°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass Bottle
Linear Formula (CH3)3COCONHCH2CO2H
Quantity 100 g
Beilstein 04, 2411
Solubility Information Solubility in water: soluble in water
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
Formula Weight 175.18
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material BOC-Glycine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-864-5

RUO – Research Use Only