missing translation for 'onlineSavingsMsg'
Learn More

BOC-D-Alanine, 99+%

Catalog No. AC275720250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275720250 25 g Glass Bottle
AC275720050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275720250 Supplier Thermo Scientific Chemicals Supplier No. 275720250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7764-95-6 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7764-95-6
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00063123
InChI Key QVHJQCGUWFKTSE-RXMQYKEDSA-N
Synonym boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
PubChem CID 637606
SMILES C[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 81°C to 84°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula (CH3)3COC(O)nHC(CH3)COOH
Specific Rotation +25° to +27° (20°C, 589nm) (c=2, CH3COOH)
Beilstein 04, 2507
IUPAC Name (2R)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Specific Rotation Condition +26° (20°C c=2,CH3COOH)
Quantity 25 g
Formula Weight 189.21
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material BOC-D-Alanine
Show More Show Less

RUO – Research Use Only