missing translation for 'onlineSavingsMsg'
Learn More

(±)-BOC-alpha-Phosphonoglycine trimethyl ester, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 439210010

Quantity 1g

 View more versions of this product

Catalog No. AC439210010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

89524-98-1
297.24
LJHAPRKTPAREGO-ZETCQYMHSA-N
397380
COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC
C10H20NO7P
MFCD02178893
methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester
methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate

Specifications

(±)-BOC-α-Phosphonoglycine trimethyl ester
96.0
Authentic
Glass bottle
MFCD02178893
methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester
COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC
297.24
297.25
89524-98-1
100.0
96% min. (GC)
C10H20NO7P
1g
LJHAPRKTPAREGO-ZETCQYMHSA-N
methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
397380
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to f

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions