missing translation for 'onlineSavingsMsg'
Learn More

BOC-beta-alanine, 99%

Catalog No. AC437651000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437651000 100 g Glass Bottle
AC437650050 5 g Glass Bottle
AC437650250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC437651000 Supplier Thermo Scientific Chemicals Supplier No. 437651000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3303-84-2 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3303-84-2
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037291
InChI Key WCFJUSRQHZPVKY-UHFFFAOYSA-N
Synonym boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
PubChem CID 76809
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCCC(O)=O

Specifications

Melting Point 77.5°C to 78.5°C
Color White
Elemental Analysis 50.02 to 51.54 % C 7.29 to 7.51 % N
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 100 g
Formula Weight 189.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material BOC-β-Alanine
Show More Show Less
EINECSNumber 221-979-2

RUO – Research Use Only