missing translation for 'onlineSavingsMsg'
Learn More

BOC-5-Aminopentanoic acid, 97%

Catalog No. p-7077318
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448240050 5 g
AC448240250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448240050 Supplier Thermo Scientific Chemicals Supplier No. 448240050

CAS: 27219-07-4 | C10H19NO4 | 217.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27219-07-4
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.26
InChI Key GFMRZAMDGJIWRB-UHFFFAOYSA-N
Synonym boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid
PubChem CID 545848
IUPAC Name 5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NCCCCC(=O)O

Specifications

Melting Point 45.0°C to 52.0°C
Boiling Point 160.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 217.26
Percent Purity 97%
Chemical Name or Material BOC-5-Aminopentanoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only