missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNAs - Sandy Loam, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| CAS | 309-00-2 |
| Type | Certified Reference Material |
| Quantity | 50 g |
| Grade | Certified Reference Material |