Learn More
Biomedical Polymers Cytology Funnels
$290.28 - $4320.55
Specifications
For Use With (Equipment) Shandon Cytospin
View More Specs
Products 5
Catalog Number Mfr. No. Quantity Type Price Quantity  
Catalog Number Mfr. No. Quantity Type Price Quantity  
23-640-321
View Documents
Biomedical Polymers
BMPCYTOS500
500/Cs. Single Funnel
Case of 500 Each for $2,237.10
Only null left
23-640-320
View Documents
Biomedical Polymers
BMPCYTOS50
50/Pk. Single Funnel
Pack of 50 Each for $290.28
Case of 10 Pack for $2,902.80
Only null left
23-640-323
View Documents
Biomedical Polymers
BMP-CYTO-DB500
500/Cs. Double Funnel
Case of 500 Each for $4,320.55
Only null left
23-640-322
View Documents
Biomedical Polymers
BMP-CYTO-DB25
25/Pk. Double Funnel
Pack of 25 Each for $318.00
Case of 10 Pack for $3,180.00
Only null left
23-640-325
View Documents
Biomedical Polymers
BMPFUNNELCLP
6/Pk. Funnel Clips
Case of 6 Each for $541.03
Only null left
Description
Description
  • For sample volumes of 0.4mL or less with single funnels and 0.25mL or less for double funnels
  • Deposits thin layer of cells in clearly defined area of microscope slide
  • Provides consistent and homogeneous cell deposition area of 6mm
  • Pre-attached filter card to facilitate absorption of excess fluid, designed for single use only
  • Funnel sealing caps included
  • Stainless steel clips available to hold funnels against slide while being centrifuged
Specifications
Specifications
Shandon Cytospin
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'