Learn More
Biomedical Polymers Cytology Funnels
Supplier:  Biomedical Polymers BMPCYTOS500
 View more versions of this product
Catalog No. 23-640-321
Only null left
Explore available promotions
Description
Description
  • For sample volumes of 0.4mL or less with single funnels and 0.25mL or less for double funnels
  • Deposits thin layer of cells in clearly defined area of microscope slide
  • Provides consistent and homogeneous cell deposition area of 6mm
  • Pre-attached filter card to facilitate absorption of excess fluid, designed for single use only
  • Funnel sealing caps included
  • Stainless steel clips available to hold funnels against slide while being centrifuged
Specifications
Specifications
Shandon Cytospin
Sealing cap
500/Cs.
Single Funnel
Product Suggestions
Product Suggestions