Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Transparent plastic material.
Dimensions: 5 x 7 in (12.7 x 17.8 cm).
Quantity: 100 per pack.
Application for Western blot imaging and handling.
Compatible with chemiluminescence and fluorescence imaging.
Not autofluorescent.
Prevents blot wrinkling and bunching.
Promotes clear, artifact-free images.
Protects delicate blots during manipulation.
Allows imaging of up to 2 blots simultaneously for efficient usage.